Anti-REEP2 Mouse Monoclonal Antibody [clone: 2C7]

Toimittaja: OriGene
ORIGCF504323EA 749 EUR
ORIGCF504323
Anti-REEP2 Mouse Monoclonal Antibody [clone: 2C7]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: REEP2
Clonality: Monoclonal
Clone: 2C7
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR