Anti-DTNB Mouse Monoclonal Antibody [clone: 3A12]

Toimittaja: OriGene
ORIGCF504326EA 749 EUR
ORIGCF504326
Anti-DTNB Mouse Monoclonal Antibody [clone: 3A12]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: DTNB
Clonality: Monoclonal
Clone: 3A12
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR