Anti-C2orf62 Mouse Monoclonal Antibody [clone: 3F11]
Toimittaja: OriGene
ORIGCF505503EA 770 EUR
ORIGCF505503
Anti-C2orf62 Mouse Monoclonal Antibody [clone: 3F11]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:4000
Type: Primary
Antigen: C2orf62
Clonality: Monoclonal
Clone: 3F11
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human