Anti-TRMT2A Mouse Monoclonal Antibody [clone: 1G8]

Toimittaja: OriGene
ORIGCF505516EA 770 EUR
ORIGCF505516
Anti-TRMT2A Mouse Monoclonal Antibody [clone: 1G8]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150

Type: Primary
Antigen: TRMT2A
Clonality: Monoclonal
Clone: 1G8
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR