Anti-CRYAA Mouse Monoclonal Antibody [clone: 2H2]
Toimittaja: OriGene
ORIGCF505529EA 749 EUR
ORIGCF505529
Anti-CRYAA Mouse Monoclonal Antibody [clone: 2H2]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000
Type: Primary
Antigen: CRYAA
Clonality: Monoclonal
Clone: 2H2
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human