Anti-GLB1 Mouse Monoclonal Antibody [clone: 1C9]

Toimittaja: OriGene
ORIGCF505544EA 749 EUR
ORIGCF505544
Anti-GLB1 Mouse Monoclonal Antibody [clone: 1C9]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:500; Immunochemistry: 1:150

Type: Primary
Antigen: GLB1
Clonality: Monoclonal
Clone: 1C9
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Dog, Human, Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR