Anti-TP53I3 Mouse Monoclonal Antibody [clone: 3E3]

Toimittaja: OriGene
ORIGCF503590EA 770 EUR
ORIGCF503590
Anti-TP53I3 Mouse Monoclonal Antibody [clone: 3E3]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: TP53I3
Clonality: Monoclonal
Clone: 3E3
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR