Anti-CAPN2 Mouse Monoclonal Antibody [clone: 1F4]

Toimittaja: OriGene
ORIGCF504345EA 764 EUR
ORIGCF504345
Anti-CAPN2 Mouse Monoclonal Antibody [clone: 1F4]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:500-2000; Flow Cytometry: 1:100

Type: Primary
Antigen: CAPN2
Clonality: Monoclonal
Clone: 1F4
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Monkey, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR