Anti-ACD Mouse Monoclonal Antibody [clone: 2B1]

Toimittaja: OriGene
ORIGCF504379EA 749 EUR
ORIGCF504379
Anti-ACD Mouse Monoclonal Antibody [clone: 2B1]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:500

Type: Primary
Antigen: ACD
Clonality: Monoclonal
Clone: 2B1
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR