Anti-PSMB9 Mouse Monoclonal Antibody [clone: 1F10]
Toimittaja: OriGene
ORIGCF504389EA 749 EUR
ORIGCF504389
Anti-PSMB9 Mouse Monoclonal Antibody [clone: 1F10]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:500-2000; Flow Cytometry: 1:100
Type: Primary
Antigen: PSMB9
Clonality: Monoclonal
Clone: 1F10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat