Anti-PSMD2 Mouse Monoclonal Antibody [clone: 1G5]

Toimittaja: OriGene
ORIGCF503198EA 749 EUR
ORIGCF503198
Anti-PSMD2 Mouse Monoclonal Antibody [clone: 1G5]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:500-2000; Immunofluorescence: 1:100

Type: Primary
Antigen: PSMD2
Clonality: Monoclonal
Clone: 1G5
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Dog, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR