Anti-TRAPPC4 Mouse Monoclonal Antibody [clone: 2G8]

Toimittaja: OriGene
ORIGCF505599EA 749 EUR
ORIGCF505599
Anti-TRAPPC4 Mouse Monoclonal Antibody [clone: 2G8]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:150; Flow Cytometry: 1:100

Type: Primary
Antigen: TRAPPC4
Clonality: Monoclonal
Clone: 2G8
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR