Anti-TMED1 Mouse Monoclonal Antibody [clone: 7D2]

Toimittaja: OriGene
ORIGCF503620EA 749 EUR
ORIGCF503620
Anti-TMED1 Mouse Monoclonal Antibody [clone: 7D2]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:500; Immunofluorescence: 1:100

Type: Primary
Antigen: TMED1
Clonality: Monoclonal
Clone: 7D2
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR