Anti-RPN1 Mouse Monoclonal Antibody [clone: 3E1]
Toimittaja: OriGene
ORIGCF503649EA 749 EUR
ORIGCF503649
Anti-RPN1 Mouse Monoclonal Antibody [clone: 3E1]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100
Type: Primary
Antigen: RPN1
Clonality: Monoclonal
Clone: 3E1
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human, Monkey, Rat