Anti-RPN1 Mouse Monoclonal Antibody [clone: 2G9]
Toimittaja: OriGene
ORIGCF503653EA 749 EUR
ORIGCF503653
Anti-RPN1 Mouse Monoclonal Antibody [clone: 2G9]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:200-1:1000; Flow Cytometry: 1:100
Type: Primary
Antigen: RPN1
Clonality: Monoclonal
Clone: 2G9
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human