Anti-NIF3L1 Mouse Monoclonal Antibody [clone: 3E1]

Toimittaja: OriGene
ORIGCF503663EA 749 EUR
ORIGCF503663
Anti-NIF3L1 Mouse Monoclonal Antibody [clone: 3E1]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: NIF3L1
Clonality: Monoclonal
Clone: 3E1
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR