Anti-CAPN9 Mouse Monoclonal Antibody [clone: 3G10]

Toimittaja: OriGene
ORIGCF503664EA 749 EUR
ORIGCF503664
Anti-CAPN9 Mouse Monoclonal Antibody [clone: 3G10]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150

Type: Primary
Antigen: CAPN9
Clonality: Monoclonal
Clone: 3G10
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Dog, Human, Monkey, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR