Anti-SPG7 Mouse Monoclonal Antibody [clone: 1C12]

Toimittaja: OriGene
ORIGCF504420EA 749 EUR
ORIGCF504420
Anti-SPG7 Mouse Monoclonal Antibody [clone: 1C12]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:500-2000; Immunofluorescence: 1:50-100; Flow Cytometry: 1:100

Type: Primary
Antigen: SPG7
Clonality: Monoclonal
Clone: 1C12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR