Anti-SCHIP1 Mouse Monoclonal Antibody [clone: 2E6]

Toimittaja: OriGene
ORIGCF504431EA 770 EUR
ORIGCF504431
Anti-SCHIP1 Mouse Monoclonal Antibody [clone: 2E6]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: SCHIP1
Clonality: Monoclonal
Clone: 2E6
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR