Anti-CNOT4 Mouse Monoclonal Antibody [clone: 3D11]

Toimittaja: OriGene
ORIGCF504447EA 749 EUR
ORIGCF504447
Anti-CNOT4 Mouse Monoclonal Antibody [clone: 3D11]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000; Flow Cytometry: 1:100

Type: Primary
Antigen: CNOT4
Clonality: Monoclonal
Clone: 3D11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR