Anti-PA2G4 Mouse Monoclonal Antibody [clone: 1D3]

Toimittaja: OriGene
ORIGCF503203EA 749 EUR
ORIGCF503203
Anti-PA2G4 Mouse Monoclonal Antibody [clone: 1D3]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: PA2G4
Clonality: Monoclonal
Clone: 1D3
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR