Anti-CBR3 Mouse Monoclonal Antibody [clone: 1G6]

Toimittaja: OriGene
ORIGCF503225EA 749 EUR
ORIGCF503225
Anti-CBR3 Mouse Monoclonal Antibody [clone: 1G6]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150

Type: Primary
Antigen: CBR3
Clonality: Monoclonal
Clone: 1G6
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human, Monkey, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR