Anti-PBX1 Mouse Monoclonal Antibody [clone: 4D1]

Toimittaja: OriGene
ORIGCF503230EA 749 EUR
ORIGCF503230
Anti-PBX1 Mouse Monoclonal Antibody [clone: 4D1]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: PBX1
Clonality: Monoclonal
Clone: 4D1
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR