Anti-SIRT5 Mouse Monoclonal Antibody [clone: 2C4]

Toimittaja: OriGene
ORIGCF503233EA 770 EUR
ORIGCF503233
Anti-SIRT5 Mouse Monoclonal Antibody [clone: 2C4]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: SIRT5
Clonality: Monoclonal
Clone: 2C4
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR