Anti-OBFC1 Mouse Monoclonal Antibody [clone: 2E4]

Toimittaja: OriGene
ORIGCF503240EA 749 EUR
ORIGCF503240
Anti-OBFC1 Mouse Monoclonal Antibody [clone: 2E4]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: OBFC1
Clonality: Monoclonal
Clone: 2E4
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR