Anti-CD44 Mouse Monoclonal Antibody [clone: 5B7]

Toimittaja: OriGene
ORIGCF506742EA 749 EUR
ORIGCF506742
Anti-CD44 Mouse Monoclonal Antibody [clone: 5B7]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:4000

Type: Primary
Antigen: CD44
Clonality: Monoclonal
Clone: 5B7
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR