Anti-CD44 Mouse Monoclonal Antibody [clone: 5E6]

Toimittaja: OriGene
ORIGCF506746EA 749 EUR
ORIGCF506746
Anti-CD44 Mouse Monoclonal Antibody [clone: 5E6]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:4000

Type: Primary
Antigen: CD44
Clonality: Monoclonal
Clone: 5E6
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR