Anti-RUNDC3A Mouse Monoclonal Antibody [clone: 3C6]

Toimittaja: OriGene
ORIGCF506769EA 749 EUR
ORIGCF506769
Anti-RUNDC3A Mouse Monoclonal Antibody [clone: 3C6]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: RUNDC3A
Clonality: Monoclonal
Clone: 3C6
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR