Anti-CD44 Mouse Monoclonal Antibody [clone: 2D9]

Toimittaja: OriGene
ORIGCF506779EA 749 EUR
ORIGCF506779
Anti-CD44 Mouse Monoclonal Antibody [clone: 2D9]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:4000; Immunochemistry: 1:150

Type: Primary
Antigen: CD44
Clonality: Monoclonal
Clone: 2D9
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR