Anti-POU5F1 Mouse Monoclonal Antibody [clone: 2B6]

Toimittaja: OriGene
ORIGCF506797EA 749 EUR
ORIGCF506797
Anti-POU5F1 Mouse Monoclonal Antibody [clone: 2B6]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:4000

Type: Primary
Antigen: POU5F1
Clonality: Monoclonal
Clone: 2B6
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR