Anti-MMP13 Mouse Monoclonal Antibody [clone: 2D8]

Toimittaja: OriGene
ORIGCF506806EA 749 EUR
ORIGCF506806
Anti-MMP13 Mouse Monoclonal Antibody [clone: 2D8]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:4000; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: MMP13
Clonality: Monoclonal
Clone: 2D8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR