Anti-FICD Mouse Monoclonal Antibody [clone: 3E3]

Toimittaja: OriGene
ORIGCF802903EA 749 EUR
ORIGCF802903
Anti-FICD Mouse Monoclonal Antibody [clone: 3E3]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:500

Type: Primary
Antigen: FICD
Clonality: Monoclonal
Clone: 3E3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR