Anti-TIPIN Mouse Monoclonal Antibody [clone: 2E2]

Toimittaja: OriGene
ORIGCF802937EA 770 EUR
ORIGCF802937
Anti-TIPIN Mouse Monoclonal Antibody [clone: 2E2]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: TIPIN
Clonality: Monoclonal
Clone: 2E2
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR