Anti-PDCD6IP Mouse Monoclonal Antibody [clone: 1A4]

Toimittaja: OriGene
ORIGCF507312EA 749 EUR
ORIGCF507312
Anti-PDCD6IP Mouse Monoclonal Antibody [clone: 1A4]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:4000; Immunochemistry: 1:150

Type: Primary
Antigen: PDCD6IP
Clonality: Monoclonal
Clone: 1A4
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR