Anti-SIRPA Mouse Monoclonal Antibody [clone: 1H1]
Toimittaja: OriGene
ORIGCF507320EA 749 EUR
ORIGCF507320
Anti-SIRPA Mouse Monoclonal Antibody [clone: 1H1]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:4000
Type: Primary
Antigen: SIRPA
Clonality: Monoclonal
Clone: 1H1
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human