Anti-ANKMY2 Mouse Monoclonal Antibody [clone: 1D8]

Toimittaja: OriGene
ORIGCF507328EA 770 EUR
ORIGCF507328
Anti-ANKMY2 Mouse Monoclonal Antibody [clone: 1D8]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:4000

Type: Primary
Antigen: ANKMY2
Clonality: Monoclonal
Clone: 1D8
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR