Anti-SIRPA Mouse Monoclonal Antibody [clone: 1B5]

Toimittaja: OriGene
ORIGCF507337EA 749 EUR
ORIGCF507337
Anti-SIRPA Mouse Monoclonal Antibody [clone: 1B5]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:4000

Type: Primary
Antigen: SIRPA
Clonality: Monoclonal
Clone: 1B5
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR