Anti-SPG7 Mouse Monoclonal Antibody [clone: 2B9]

Toimittaja: OriGene
ORIGCF800005EA 749 EUR
ORIGCF800005
Anti-SPG7 Mouse Monoclonal Antibody [clone: 2B9]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100

Type: Primary
Antigen: SPG7
Clonality: Monoclonal
Clone: 2B9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR