Anti-AMACR Mouse Monoclonal Antibody [clone: 5G8]

Toimittaja: OriGene
ORIGCF800837EA 749 EUR
ORIGCF800837
Anti-AMACR Mouse Monoclonal Antibody [clone: 5G8]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150

Type: Primary
Antigen: AMACR
Clonality: Monoclonal
Clone: 5G8
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR