Anti-ABCB1 Mouse Monoclonal Antibody [clone: 2G7]

Toimittaja: OriGene
ORIGCF800920EA 749 EUR
ORIGCF800920
Anti-ABCB1 Mouse Monoclonal Antibody [clone: 2G7]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: ABCB1
Clonality: Monoclonal
Clone: 2G7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR