Anti-LGALS3 Mouse Monoclonal Antibody [clone: 1C7]

Toimittaja: OriGene
ORIGCF801621EA 749 EUR
ORIGCF801621
Anti-LGALS3 Mouse Monoclonal Antibody [clone: 1C7]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150

Type: Primary
Antigen: LGALS3
Clonality: Monoclonal
Clone: 1C7
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR