Anti-KRAS Mouse Monoclonal Antibody [clone: 2C1]

Toimittaja: OriGene
ORIGCF801672EA 749 EUR
ORIGCF801672
Anti-KRAS Mouse Monoclonal Antibody [clone: 2C1]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: KRAS
Clonality: Monoclonal
Clone: 2C1
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR