Anti-CD68 Mouse Monoclonal Antibody [clone: 6D3]

Toimittaja: OriGene
ORIGCF802966EA 749 EUR
ORIGCF802966
Anti-CD68 Mouse Monoclonal Antibody [clone: 6D3]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: CD68
Clonality: Monoclonal
Clone: 6D3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR