Anti-FBXW7 Mouse Monoclonal Antibody [clone: 4E7]

Toimittaja: OriGene
ORIGCF802970EA 749 EUR
ORIGCF802970
Anti-FBXW7 Mouse Monoclonal Antibody [clone: 4E7]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: FBXW7
Clonality: Monoclonal
Clone: 4E7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR