Anti-SPG7 Mouse Monoclonal Antibody [clone: 7D10]

Toimittaja: OriGene
ORIGCF800012EA 764 EUR
ORIGCF800012
Anti-SPG7 Mouse Monoclonal Antibody [clone: 7D10]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: SPG7
Clonality: Monoclonal
Clone: 7D10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR