Anti-CRABP2 Mouse Monoclonal Antibody [clone: 3C11]

Toimittaja: OriGene
ORIGCF800027EA 749 EUR
ORIGCF800027
Anti-CRABP2 Mouse Monoclonal Antibody [clone: 3C11]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:500-2000

Type: Primary
Antigen: CRABP2
Clonality: Monoclonal
Clone: 3C11
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR