Anti-DPPA4 Mouse Monoclonal Antibody [clone: 1E1]

Toimittaja: OriGene
ORIGCF800032EA 770 EUR
ORIGCF800032
Anti-DPPA4 Mouse Monoclonal Antibody [clone: 1E1]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000; Flow Cytometry: 1:100

Type: Primary
Antigen: DPPA4
Clonality: Monoclonal
Clone: 1E1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR