Anti-USP12 Mouse Monoclonal Antibody [clone: 2A6]

Toimittaja: OriGene
ORIGCF800037EA 749 EUR
ORIGCF800037
Anti-USP12 Mouse Monoclonal Antibody [clone: 2A6]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: USP12
Clonality: Monoclonal
Clone: 2A6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR