Anti-CNOT4 Mouse Monoclonal Antibody [clone: 1F10]

Toimittaja: OriGene
ORIGCF800048EA 749 EUR
ORIGCF800048
Anti-CNOT4 Mouse Monoclonal Antibody [clone: 1F10]
Vasta-aineet
Recommended Dilutions: Immunochemistry: 1:150

Type: Primary
Antigen: CNOT4
Clonality: Monoclonal
Clone: 1F10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR