Anti-S100A6 Mouse Monoclonal Antibody [clone: 6F8]

Toimittaja: OriGene
ORIGCF800941EA 749 EUR
ORIGCF800941
Anti-S100A6 Mouse Monoclonal Antibody [clone: 6F8]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: S100A6
Clonality: Monoclonal
Clone: 6F8
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR